Assay Details
Target Gene Details
Entrez Gene ID: | 67338 |
Gene Name: | ring finger and FYVE like domain containing protein |
Gene Aliases: |
1700051E09Rik, 4930516L10Rik, BG080975, Carp2 |
Location: |
Chr.11:82803819-82871239 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rffl | NM_001007465.3 | NP_001007466.1 | ||
| NM_001164569.1 | NP_001158041.1 | |||
| NM_001164570.1 | NP_001158042.1 | |||
| NM_001164571.1 | NP_001158043.1 | |||
| NM_026097.3 | NP_080373.1 | |||
| AF434814.1 | AAL30769.1 | |||
| AK006757.1 | ||||
| AK007189.1 | BAB24891.1 | |||
| AK015806.1 | BAB29984.1 | |||
| AK028095.1 | BAC25744.1 | |||
| AK128983.1 | BAC87665.1 | |||
| AK150213.1 | BAE29383.1 | |||
| AK150541.1 | BAE29644.1 | |||
| AK159442.1 | ||||
| AK169393.1 | BAE41140.1 | |||
| AK169988.1 | ||||
| BC118517.1 |
More Information
Set Membership: |
|

Genomic Map