Assay Details
Target Gene Details
Entrez Gene ID: | 109342 |
Gene Name: | solute carrier family 5 (sodium/glucose cotransporter), member 10 |
Gene Aliases: |
C330021F16Rik, SGLT5 |
Location: |
Chr.11:61672782-61720811 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc5a10 | NM_001033227.2 | 13 | 1531 | NP_001028399.1 |
| AK082798.1 | 2 | 187 | ||
| AK144033.1 | 13 | 1532 | BAE25665.1 | |
| AK165337.1 | 12 | 1327 | BAE38140.1 | |
| BC141325.1 | 12 | 1324 |
More Information
Set Membership: |
|

Genomic Map