Assay Details
Target Gene Details
Entrez Gene ID: | 22123 |
Gene Name: | proteasome (prosome, macropain) 26S subunit, non-ATPase, 3 |
Gene Aliases: |
AI255837, AntP91a, D11Bwg1349e, Psd3, Tstap91a |
Location: |
Chr.11:98682554-98695978 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psmd3 | NM_009439.1 | 4 | 781 | NP_033465.1 |
| AK031366.1 | 4 | 771 | ||
| AK077252.1 | 4 | 730 | BAC36709.1 | |
| AK089933.1 | 4 | 739 | ||
| AK145467.1 | 4 | 771 | BAE26453.1 | |
| AK145488.1 | 4 | 770 | BAE26466.1 | |
| AK153505.1 | 4 | 698 | BAE32050.1 | |
| AK164333.1 | 4 | 745 | ||
| AK164593.1 | 4 | 770 | BAE37842.1 | |
| AK165520.1 | 4 | 716 | BAE38235.1 | |
| AK166683.1 | 4 | 741 | BAE38942.1 | |
| AK166714.1 | 4 | 769 | BAE38965.1 | |
| AK167005.1 | 4 | 756 | ||
| AK167285.1 | 4 | 736 | BAE39391.1 | |
| AK169690.1 | 4 | 770 | BAE41307.1 | |
| AK180273.1 | 2 | 157 | ||
| BC003197.1 | 4 | 781 | AAH03197.1 |
More Information
Set Membership: |
|

Genomic Map