Assay Details
Target Gene Details
Entrez Gene ID: | 67815 |
Gene Name: | SEC14-like 2 (S. cerevisiae) |
Gene Aliases: |
1300013M05Rik, Spf, TAP |
Location: |
Chr.11:4097039-4118729 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sec14l2 | NM_144520.2 | NP_653103.1 | ||
| BC005759.1 | AAH05759.1 |

Genomic Map