Assay Details
Target Gene Details
Entrez Gene ID: | 380683 |
Gene Name: | SEC14-like 3 (S. cerevisiae) |
Gene Aliases: |
1110069O07Rik |
Location: |
Chr.11:4064853-4078990 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sec14l3 | NM_001029937.2 | NP_001025108.1 | ||
| AK136060.1 | BAE22802.1 | |||
| BC138982.1 | ||||
| BC138985.1 |

Genomic Map