Assay Details
Target Gene Details
Entrez Gene ID: | 67863 |
Gene Name: | solute carrier family 25 (mitochondrial carrier oxoglutarate carrier), member 11 |
Gene Aliases: |
2310022P18Rik, 2oxoc |
Location: |
Chr.11:70644019-70647423 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a11 | NM_024211.3 | 3 | 578 | NP_077173.1 |
| AI931249.1 | ||||
| AK003957.1 | 2 | 217 | BAB23092.1 | |
| AK009487.1 | 3 | 524 | BAB26319.1 | |
| AK009824.1 | 3 | 478 | BAB26524.1 | |
| AK168951.1 | 3 | 503 | BAE40757.1 | |
| AK181540.1 | 2 | 260 | ||
| AK187524.1 | 2 | 260 | ||
| AK202646.1 | 2 | 260 | ||
| AK212146.1 | 2 | 250 | ||
| AK213879.1 | 2 | 260 | ||
| AK215545.1 | 2 | 250 | ||
| BC003455.1 | 3 | 554 | AAH03455.1 | |
| BC019631.1 | 3 | 532 | AAH19631.1 |
More Information
Set Membership: |
|

Genomic Map