Assay Details
Target Gene Details
Entrez Gene ID: | 67283 |
Gene Name: | solute carrier family 25 (mitochondrial thiamine pyrophosphate carrier), member 19 |
Gene Aliases: |
2900089E13Rik, DNC, MUP1, TPC |
Location: |
Chr.11:115614170-115628295 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a19 | NM_001252384.1 | 1 | 112 | NP_001239313.1 |
| NM_001252394.1 | 1 | 112 | NP_001239323.1 | |
| NM_001252395.1 | 1 | 112 | NP_001239324.1 | |
| NM_001252396.1 | 1 | 112 | NP_001239325.1 | |
| NM_026071.3 | 1 | 112 | NP_080347.1 | |
| BY276424.1 | 1 | 114 |
More Information
Set Membership: |
|

Genomic Map