Assay Details
Target Gene Details
Entrez Gene ID: | 67283 |
Gene Name: | solute carrier family 25 (mitochondrial thiamine pyrophosphate carrier), member 19 |
Gene Aliases: |
2900089E13Rik, DNC, MUP1, TPC |
Location: |
Chr.11:115614170-115628295 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a19 | NM_001252384.1 | NP_001239313.1 | ||
| NM_001252394.1 | NP_001239323.1 | |||
| NM_001252395.1 | NP_001239324.1 | |||
| NM_001252396.1 | NP_001239325.1 | |||
| NM_026071.3 | NP_080347.1 | |||
| AK005710.1 | BAB24199.1 | |||
| AK168145.1 | ||||
| AK172622.1 | BAE43100.1 | |||
| AK194051.1 | ||||
| BC018167.1 | AAH18167.1 | |||
| BC046767.1 | AAH46767.1 | |||
| BI736130.1 | ||||
| CF730205.1 | ||||
| CX241396.1 |
More Information
Set Membership: |
|

Genomic Map