Assay Details
Target Gene Details
Entrez Gene ID: | 26941 |
Gene Name: | solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 1 |
Gene Aliases: |
EBP-50, NHE-RF, NHERF-1, NHERF1 |
Location: |
Chr.11:115163312-115181183 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc9a3r1 | NM_012030.2 | NP_036160.1 | ||
| AK085210.1 | ||||
| AK153558.1 | BAE32094.1 | |||
| AK168894.1 | BAE40711.1 | |||
| AK177064.1 | ||||
| AK188344.1 | ||||
| AK205456.1 | ||||
| AK209994.1 | ||||
| AK216315.1 | ||||
| BC085141.1 | AAH85141.1 |

Genomic Map