Assay Details
Target Gene Details
Entrez Gene ID: | 320655 |
Gene Name: | post-GPI attachment to proteins 3 |
Gene Aliases: |
CAB2, D430035D22Rik, Perld1 |
Location: |
Chr.11:98388672-98400539 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pgap3 | NM_001033537.2 | 7 | 855 | NP_001028709.2 |
| AK052486.1 | 7 | 772 | ||
| AK078974.1 | 7 | 781 | ||
| AK170817.1 | 7 | 770 | BAE42049.1 | |
| AK187756.1 | ||||
| BC030368.1 | 5 | 475 | AAH30368.1 |

Genomic Map