Assay Details
Target Gene Details
Entrez Gene ID: | 54351 |
Gene Name: | elongator acetyltransferase complex subunit 5 |
Gene Aliases: |
Derp6, Rai12 |
Location: |
Chr.11:69968224-69981294 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elp5 | NM_001253700.1 | NP_001240629.1 | ||
| NM_018740.2 | NP_061210.2 | |||
| AA646690.1 | ||||
| AF004107.1 | AAC16252.1 | |||
| AK011130.1 | 1 | 144 | ||
| AK152582.1 | BAE31332.1 | |||
| BC003752.1 | AAH03752.1 | |||
| BY292587.1 | 1 | 134 |
More Information
Set Membership: |
|

Genomic Map