Assay Details
Target Gene Details
Entrez Gene ID: | 54351 |
Gene Name: | elongator acetyltransferase complex subunit 5 |
Gene Aliases: |
Derp6, Rai12 |
Location: |
Chr.11:69968224-69981294 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elp5 | NM_001253700.1 | NP_001240629.1 | ||
| NM_018740.2 | NP_061210.2 | |||
| AF004107.1 | AAC16252.1 | |||
| AK011130.1 | ||||
| AK013662.1 | BAB28944.1 | |||
| AK152582.1 | BAE31332.1 | |||
| AK207946.1 | ||||
| BC003752.1 | AAH03752.1 | |||
| BE862527.1 | ||||
| CD741073.1 | ||||
| CD776986.1 |

Genomic Map