Assay Details
Target Gene Details
Entrez Gene ID: | 268448 |
Gene Name: | PHD finger protein 12 |
Gene Aliases: |
2410142K10Rik, F630045O13, PF1, mKIAA1523 |
Location: |
Chr.11:77982754-78030539 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phf12 | NM_174852.3 | NP_777277.2 | ||
| AK089226.1 | ||||
| AK129383.1 | BAC98193.2 | |||
| AK141796.1 | BAE24837.1 | |||
| AK155354.1 | BAE33212.1 | |||
| AK170155.1 | BAE41602.1 | |||
| AK170187.1 | BAE41622.1 | |||
| BC043080.1 | AAH43080.1 |
More Information
Set Membership: |
|

Genomic Map