Assay Details
Target Gene Details
Entrez Gene ID: | 237636 |
Gene Name: | NPC1-like 1 |
Gene Aliases: |
9130221N23Rik, Gm243 |
Location: |
Chr.11:6211011-6230245 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npc1l1 | NM_207242.2 | NP_997125.2 | ||
| AK078947.1 | ||||
| AY437866.1 | AAR97887.1 | |||
| BC131788.1 | ||||
| BC131789.1 |

Genomic Map