Assay Details
Target Gene Details
Entrez Gene ID: | 18293 |
Gene Name: | oxoglutarate (alpha-ketoglutarate) dehydrogenase (lipoamide) |
Gene Aliases: |
2210403E04Rik, 2210412K19Rik, AA409584, d1401, mKIAA4192 |
Location: |
Chr.11:6291593-6359094 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ogdh | NM_001252282.1 | 16 | 2099 | NP_001239211.1 |
| NM_001252283.1 | 16 | 2087 | NP_001239212.1 | |
| NM_001252287.1 | 15 | 2130 | NP_001239216.1 | |
| NM_001252288.1 | 15 | 2102 | NP_001239217.1 | |
| NM_010956.4 | 15 | 2054 | NP_035086.2 | |
| AK147289.1 | 15 | 2055 | BAE27824.1 | |
| AK150009.1 | 15 | 2020 | BAE29234.1 | |
| AK169286.1 | 15 | 2046 | BAE41044.1 | |
| AK220536.1 | 16 | 2061 | BAD90530.1 | |
| BC013670.1 | 15 | 1988 | AAH13670.1 | |
| BC049104.1 | 15 | 2055 | AAH49104.1 | |
| BC057354.1 | 16 | 2015 | AAH57354.1 | |
| BC061232.1 | AAH61232.1 |
More Information
Set Membership: |
|

Genomic Map