Assay Details
Target Gene Details
Entrez Gene ID: | 18293 |
Gene Name: | oxoglutarate (alpha-ketoglutarate) dehydrogenase (lipoamide) |
Gene Aliases: |
2210403E04Rik, 2210412K19Rik, AA409584, d1401, mKIAA4192 |
Location: |
Chr.11:6291593-6359094 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ogdh | NM_001252282.1 | 2 | 257 | NP_001239211.1 |
| NM_001252283.1 | 2 | 257 | NP_001239212.1 | |
| NM_001252287.1 | 2 | 333 | NP_001239216.1 | |
| NM_001252288.1 | 2 | 317 | NP_001239217.1 | |
| NM_010956.4 | 2 | 257 | NP_035086.2 | |
| AK008811.1 | 2 | 213 | ||
| AK147289.1 | 2 | 258 | BAE27824.1 | |
| AK150009.1 | 2 | 223 | BAE29234.1 | |
| AK169286.1 | 2 | 249 | BAE41044.1 | |
| AK195097.1 | 1 | 37 | ||
| AK201619.1 | ||||
| AK202346.1 | 1 | 37 | ||
| AK204071.1 | ||||
| AK213993.1 | 1 | 37 | ||
| AK220536.1 | 2 | 219 | BAD90530.1 | |
| BC013670.1 | 2 | 191 | AAH13670.1 | |
| BC049104.1 | 2 | 270 | AAH49104.1 | |
| BC057354.1 | 2 | 186 | AAH57354.1 | |
| BC061232.1 | 2 | 213 | AAH61232.1 | |
| BY768178.1 | 2 | 250 | ||
| BY782774.1 | ||||
| BY785860.1 |
More Information
Set Membership: |
|

Genomic Map