Assay Details
Target Gene Details
Entrez Gene ID: | 18559 |
Gene Name: | phosphatidylcholine transfer protein |
Gene Aliases: |
PC-TP, StarD2 |
Location: |
Chr.11:89983417-90002914 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pctp | NM_008796.2 | NP_032822.2 | ||
| AF151639.1 | AAF02537.1 | |||
| BY144797.1 | ||||
| Z50024.1 | CAA90328.1 |

Genomic Map