Assay Details
Target Gene Details
Entrez Gene ID: | 57230 |
Gene Name: | SAP30 binding protein |
Gene Aliases: |
2700016D05Rik, Hcngp |
Location: |
Chr.11:115933578-115965534 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sap30bp | NM_020483.3 | NP_065229.2 | ||
| AK012245.1 | ||||
| AK147869.1 | BAE28192.1 | |||
| AK183667.1 | ||||
| AK210436.1 | ||||
| BC021757.1 | AAH21757.1 | |||
| X68061.1 | CAA48198.1 |
More Information
Set Membership: |
|

Genomic Map