Assay Details
Target Gene Details
Entrez Gene ID: | 104910 |
Gene Name: | solute carrier family 25, member 47 |
Gene Aliases: |
AI132487, AI876593, HDMCP |
Location: |
Chr.12:108849823-108856815 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a47 |

Genomic Map