Assay Details
Target Gene Details
Entrez Gene ID: | 216965 |
Gene Name: | TAO kinase 1 |
Gene Aliases: |
2810468K05Rik, AU020252, D130018F14Rik, Map3k16, Markk, Psk2, mKIAA1361 |
Location: |
Chr.11:77529162-77607860 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Taok1 | NM_144825.2 | 17 | 2723 | NP_659074.2 |
| AK173156.1 | 1 | 57 | BAD32434.1 | |
| BC016522.1 | 5 | 717 | AAH16522.1 | |
| BC151012.1 | 17 | 2517 | ||
| BC151015.1 | 17 | 2517 | ||
| BC151016.1 | 17 | 2516 | ||
| CN717576.1 | 4 | 531 |
More Information
Set Membership: |
|

Genomic Map