Assay Details
Target Gene Details
Entrez Gene ID: | 83560 |
Gene Name: | testis expressed gene 14 |
Gene Aliases: |
C85585 |
Location: |
Chr.11:87405050-87555823 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Intron 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tex14 | NM_001199293.1 | NP_001186222.1 | ||
| NM_031386.2 | NP_113563.2 | |||
| AK139808.1 | BAE24144.1 |
More Information
Set Membership: |
|

Genomic Map