Assay Details
Target Gene Details
Entrez Gene ID: | 76281 |
Gene Name: | Tax1 (human T cell leukemia virus type I) binding protein 3 |
Gene Aliases: |
1300011C24Rik, TIP-1 |
Location: |
Chr.11:73177083-73182046 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tax1bp3 | NM_029564.2 | NP_083840.1 | ||
| AK004963.1 | BAB23703.1 | |||
| AK150816.1 | BAE29879.1 | |||
| AK168533.1 | BAE40411.1 | |||
| AK170485.1 | BAE41827.1 | |||
| BC008166.1 | AAH08166.1 | |||
| BC094314.1 | AAH94314.1 |

Genomic Map