Assay Details
Target Gene Details
Entrez Gene ID: | 83560 |
Gene Name: | testis expressed gene 14 |
Gene Aliases: |
C85585 |
Location: |
Chr.11:87405050-87555823 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tex14 | NM_001199293.1 | 3 | 323 | NP_001186222.1 |
NM_031386.2 | 3 | 304 | NP_113563.2 | |
AY193717.1 | 3 | 304 | AAN86761.1 | |
AY193718.1 | 3 | 323 | AAN86762.1 |
