Assay Details
Target Gene Details
Entrez Gene ID: | 327959 |
Gene Name: | XIAP associated factor 1 |
Gene Aliases: |
Fbox39 |
Location: |
Chr.11:72301436-72313733 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xaf1 | NM_001037713.4 | NP_001032802.2 | ||
| NM_001291153.1 | NP_001278082.1 | |||
| AK046736.1 | ||||
| AK140315.1 | BAE24328.1 | |||
| BY726623.1 | ||||
| EL957551.1 |

Genomic Map