Assay Details
Target Gene Details
Entrez Gene ID: | 20892 |
Gene Name: | stimulated by retinoic acid 13 |
Gene Aliases: |
- |
Location: |
Chr.11:120710941-120713903 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stra13 | NM_016665.2 | NP_057874.2 | ||
| AK076181.1 | BAC36237.1 | |||
| AK080500.1 | BAC37933.1 | |||
| AK170126.1 | BAE41581.1 | |||
| CA320983.1 | ||||
| CJ236047.1 | ||||
| U95003.1 | AAB53635.1 | |||
| U95004.1 | AAB53636.1 | |||
| U95005.1 | AAB53637.1 |
More Information
Set Membership: |
|

Genomic Map