Assay Details
Target Gene Details
Entrez Gene ID: | 27364 |
Gene Name: | serine racemase |
Gene Aliases: |
Srs |
Location: |
Chr.11:74905047-74925798 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srr | NM_001163311.1 | 1 | 45 | NP_001156783.1 |
| NM_013761.4 | 1 | 45 | NP_038789.1 | |
| AB232340.1 | BAE19920.1 | |||
| AB232342.1 | 1 | 45 | BAE19922.1 | |
| AK157122.1 | 1 | 196 | BAE33968.1 |
More Information
Set Membership: |
|

Genomic Map