Assay Details
Target Gene Details
Entrez Gene ID: | 27364 |
Gene Name: | serine racemase |
Gene Aliases: |
Srs |
Location: |
Chr.11:74905047-74925798 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srr | NM_001163311.1 | 2 | 303 | NP_001156783.1 |
| NM_013761.4 | 3 | 529 | NP_038789.1 | |
| AB232340.1 | 3 | 481 | BAE19920.1 | |
| AB232341.1 | 3 | 469 | BAE19921.1 | |
| AB232342.1 | 2 | 303 | BAE19922.1 | |
| AB232343.1 | 1 | 301 | BAE19923.1 | |
| AB235396.1 | 2 | 145 | BAE44529.1 | |
| AF148321.1 | 2 | 184 | AAF08701.1 | |
| AK017235.1 | 3 | 409 | ||
| AK028034.1 | 2 | 186 | BAC25712.1 | |
| AK031687.1 | 3 | 401 | BAC27514.1 | |
| AK043738.1 | 3 | 435 | BAC31637.1 | |
| AK043819.1 | 2 | 209 | ||
| AK079343.1 | ||||
| AK080830.1 | 3 | 408 | ||
| AK157122.1 | 2 | 454 | BAE33968.1 | |
| AK170096.1 | 3 | 415 | BAE41561.1 | |
| AK170471.1 | 3 | 415 | ||
| BC011164.1 | 3 | 406 | AAH11164.1 |
More Information
Set Membership: |
|

Genomic Map