Assay Details
Target Gene Details
Entrez Gene ID: | 237860 |
Gene Name: | slingshot homolog 2 (Drosophila) |
Gene Aliases: |
SSH-2, SSH-2L, mSSH-2L |
Location: |
Chr.11:77216169-77460220 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ssh2 | NM_177710.4 | NP_808378.2 | ||
| AB099288.1 | BAC97811.1 | |||
| BC141392.1 | ||||
| BC141393.1 |
More Information
Set Membership: |
|

Genomic Map