Assay Details
Target Gene Details
Entrez Gene ID: | 66048 |
Gene Name: | ER membrane protein complex subunit 6 |
Gene Aliases: |
0610009E20Rik, 0610025L18Rik, Tmem93 |
Location: |
Chr.11:73175503-73177042 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Emc6 | NM_001168470.1 | NP_001161942.1 | ||
| NM_025318.3 | NP_079594.1 | |||
| AK002385.1 | BAB22059.1 | |||
| AK003198.1 | BAB22636.1 | |||
| AK004149.1 | BAB23194.1 | |||
| AK012769.1 | BAB28457.1 | |||
| BC022104.1 | AAH22104.1 |
Target Gene Details
Entrez Gene ID: | 76281 |
Gene Name: | Tax1 (human T cell leukemia virus type I) binding protein 3 |
Gene Aliases: |
1300011C24Rik, TIP-1 |
Location: |
Chr.11:73177083-73182046 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tax1bp3 | NM_029564.2 | NP_083840.1 | ||
| AK004963.1 | BAB23703.1 |

Genomic Map