Assay Details
Target Gene Details
Entrez Gene ID: | 100503361 |
Gene Name: | transmembrane protein 95 |
Gene Aliases: |
432576 |
Location: |
Chr.11:69876054-69878173 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem95 | NM_001195710.1 | 5 | 373 | NP_001182639.1 |
| BF018485.1 | 5 | 320 |

Genomic Map