Assay Details
Target Gene Details
Entrez Gene ID: | 21973 |
Gene Name: | topoisomerase (DNA) II alpha |
Gene Aliases: |
Top-2 |
Location: |
Chr.11:98992947-99024206 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 26 - Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Top2a | NM_011623.2 | NP_035753.2 | ||
| AK033321.1 | BAC28232.1 | |||
| AK138079.1 | BAE23548.1 | |||
| AK161381.1 | BAE36361.1 | |||
| AK169723.1 | ||||
| BC094326.1 | AAH94326.1 | |||
| D12513.1 | BAA02076.1 |
More Information
Set Membership: |
|

Genomic Map