Assay Details
Target Gene Details
Entrez Gene ID: | 66827 |
Gene Name: | tetratricopeptide repeat domain 1 |
Gene Aliases: |
4833412C19Rik, C79328, TPR1 |
Location: |
Chr.11:43730006-43747973 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc1 | NM_133795.1 | NP_598556.1 | ||
| AK077715.1 | BAC36975.1 | |||
| AK166431.1 | BAE38772.1 | |||
| AK170032.1 | ||||
| AK170230.1 | BAE41649.1 | |||
| BC010236.1 | AAH10236.1 |
More Information
Set Membership: |
|

Genomic Map