Assay Details
Target Gene Details
Entrez Gene ID: | 74407 |
Gene Name: | tetratricopeptide repeat domain 25 |
Gene Aliases: |
4933404O19Rik |
Location: |
Chr.11:100545613-100572566 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc25 | NM_028918.2 | NP_083194.2 | ||
| AK016657.1 | BAB30363.1 | |||
| AK161541.1 | ||||
| AK162855.1 | ||||
| BC050805.1 | ||||
| CA463990.1 |

Genomic Map