Assay Details
Target Gene Details
Entrez Gene ID: | 74407 |
Gene Name: | tetratricopeptide repeat domain 25 |
Gene Aliases: |
4933404O19Rik |
Location: |
Chr.11:100545613-100572566 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc25 | NM_028918.2 | 9 | 1404 | NP_083194.2 |
| AK016657.1 | 9 | 1335 | BAB30363.1 | |
| AK136690.1 | 3 | 249 | BAE23101.1 | |
| AK161541.1 | 9 | 1343 | ||
| AK162855.1 | 9 | 1344 | ||
| BC050805.1 | 9 | 1426 |

Genomic Map