Assay Details
Target Gene Details
Entrez Gene ID: | 238161 |
Gene Name: | A kinase (PRKA) anchor protein 6 |
Gene Aliases: |
AI482140, Akapalpha, Akapbeta, PRKA |
Location: |
Chr.12:52699016-53155568 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akap6 | NM_198111.2 | 12 | 3603 | NP_932779.2 |
| AK147248.1 | 12 | 3604 | BAE27795.1 | |
| AK147371.1 | 12 | 3604 | BAE27869.1 | |
| AK162929.1 | BAE37120.1 | |||
| BC057114.1 | 11 | 3553 | AAH57114.1 | |
| DQ233645.1 | 12 | 3659 | ABB60211.1 | |
| DQ233646.1 | 11 | 3453 | ABB60212.1 |

Genomic Map