Assay Details
Target Gene Details
Entrez Gene ID: | 21975 |
Gene Name: | topoisomerase (DNA) III alpha |
Gene Aliases: |
- |
Location: |
Chr.11:60739473-60777365 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Top3a | NM_009410.2 | 6 | 877 | NP_033436.1 |
| AB006074.1 | 6 | 878 | BAA25662.1 | |
| AK029987.1 | 3 | 270 | BAC26718.1 | |
| AK037640.1 | 6 | 778 | BAE20515.1 | |
| AK164443.1 | 6 | 778 | BAE37792.1 | |
| AK170291.1 | 7 | 684 | BAE41691.1 | |
| AK171286.1 | 7 | 684 | BAE42369.1 | |
| BC099689.1 | 6 | 692 | AAH99689.1 |
More Information
Set Membership: |
|

Genomic Map