Assay Details
Target Gene Details
Entrez Gene ID: | 276891 |
Gene Name: | T cell immunoglobulin and mucin domain containing 4 |
Gene Aliases: |
B430010N18Rik, TIM-4, Tim4 |
Location: |
Chr.11:46810786-46844333 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Timd4 | NM_178759.4 | NP_848874.3 | ||
| AK046560.1 | BAC32787.1 | |||
| AK079787.1 | BAC37750.1 | |||
| AK080042.1 | BAC37813.1 | |||
| AK080212.1 | BAC37848.1 | |||
| AY376716.1 | AAR23483.1 | |||
| AY376717.1 | AAR23484.1 |

Genomic Map