Assay Details
Target Gene Details
Entrez Gene ID: | 104776 |
Gene Name: | aldehyde dehydrogenase family 6, subfamily A1 |
Gene Aliases: |
1110038I05Rik, AI314632, Mmsdh |
Location: |
Chr.12:84430717-84450950 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aldh6a1 | NM_134042.2 | 8 | 998 | NP_598803.1 |
| AF297860.1 | 8 | 998 | AAG44988.1 | |
| AK033587.1 | 8 | 1025 | BAC28375.1 | |
| AK082790.1 | 8 | 1052 | ||
| AK128924.1 | 8 | 1030 | ||
| AK147146.1 | 8 | 1052 | BAE27715.1 | |
| AK155814.1 | 8 | 1049 | BAE33445.1 | |
| AK169915.1 | 8 | 1038 | BAE41455.1 | |
| AK170305.1 | 8 | 1051 | BAE41702.1 | |
| AK171581.1 | 8 | 1052 | BAE42539.1 | |
| AK171896.1 | 8 | 1041 | BAE42726.1 | |
| AK184313.1 | 2 | 193 | ||
| AK211862.1 | 2 | 193 | ||
| AK215486.1 | 2 | 193 | ||
| BC031148.1 | 8 | 1123 | AAH31148.1 | |
| BC033440.1 | 7 | 1021 | AAH33440.1 |
Target Gene Details
Entrez Gene ID: | 72873 |
Gene Name: | coiled-coil domain containing 176 |
Gene Aliases: |
2900006K08Rik, AI427784 |
Location: |
Chr.12:84409068-84441763 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc176 |
More Information
Set Membership: |
|

Genomic Map