Assay Details
Target Gene Details
Entrez Gene ID: | 59045 |
Gene Name: | START domain containing 3 |
Gene Aliases: |
Mln64, es64 |
Location: |
Chr.11:98358367-98381112 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stard3 | NM_021547.3 | NP_067522.1 | ||
| AK041538.1 | BAC30977.1 | |||
| AK150166.1 | ||||
| AK150254.1 | ||||
| AK155124.1 | BAE33063.1 | |||
| AK160480.1 | ||||
| AK205267.1 | ||||
| BC003313.1 | AAH03313.1 | |||
| X82457.1 | CAA57834.1 |

Genomic Map