Assay Details
Target Gene Details
Entrez Gene ID: | 380709 |
Gene Name: | spermatogenesis associated 22 |
Gene Aliases: |
Gm882 |
Location: |
Chr.11:73326537-73346434 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spata22 | NM_001045531.1 | 3 | 209 | NP_001038996.1 |
| BC147586.1 | 3 | 191 | ||
| BC147607.1 | 3 | 191 |

Genomic Map