Assay Details
Target Gene Details
Entrez Gene ID: | 217069 |
Gene Name: | tripartite motif-containing 25 |
Gene Aliases: |
AA960166, AL022677, EFP, Zfp147 |
Location: |
Chr.11:88999403-89020293 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trim25 | NM_009546.2 | NP_033572.2 | ||
| AK154533.1 | BAE32658.1 | |||
| AK160898.1 | BAE36077.1 | |||
| AK169562.1 | BAE41230.1 | |||
| AK171744.1 | BAE42646.1 | |||
| BC006908.1 | AAH06908.1 | |||
| BC034276.1 | AAH34276.1 | |||
| D63902.1 | BAA09941.1 |

Genomic Map