Assay Details
Target Gene Details
Entrez Gene ID: | 83813 |
Gene Name: | tyrosine kinase, non-receptor, 1 |
Gene Aliases: |
Kos1, Tnk1a, Tnk1b |
Location: |
Chr.11:69850563-69858730 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnk1 | NM_031880.3 | 3 | 193 | NP_114086.3 |
| AF307746.1 | 3 | 193 | AAL09413.1 | |
| AF332512.1 | AAK35164.1 | |||
| AK076220.1 | 3 | 194 | ||
| AK167292.1 | 3 | 194 | BAE39397.1 | |
| BC029623.1 | 2 | 180 | AAH29623.1 | |
| BC055303.1 | 2 | 97 | AAH55303.1 |

Genomic Map