Assay Details
Target Gene Details
Entrez Gene ID: | 217342 |
Gene Name: | ubiquitin-conjugating enzyme E2O |
Gene Aliases: |
9630022H21, B230113M03Rik, E2-230K, mKIAA1734 |
Location: |
Chr.11:116537740-116581476 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ube2o | NM_173755.3 | 16 | 3071 | NP_776116.2 |
| AK035970.1 | 1 | 128 | ||
| AK045398.1 | 3 | 441 | BAC32345.1 | |
| AK129431.1 | 16 | 3071 | BAC98241.1 | |
| BC059193.1 | 16 | 2696 | AAH59193.1 |

Genomic Map