Assay Details
Target Gene Details
Entrez Gene ID: | 268470 |
Gene Name: | ubiquitin-conjugating enzyme E2Z (putative) |
Gene Aliases: |
AW049199, C030047H17Rik, D11Moh35 |
Location: |
Chr.11:96047431-96065364 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ube2z | NM_172300.3 | 3 | 541 | NP_758504.3 |
| AK005524.1 | 1 | 92 | BAB24097.1 | |
| AK077793.1 | 3 | 540 | BAC37014.1 | |
| AK149770.1 | 3 | 541 | BAE29074.1 | |
| AK152529.1 | 3 | 255 | ||
| AK153328.1 | 3 | 272 | BAE31907.1 | |
| AK168942.1 | 3 | 234 | BAE40750.1 | |
| AK203763.1 | 2 | 130 | ||
| BC054412.1 | 3 | 244 | AAH54412.1 | |
| CA317477.1 | 3 | 565 |

Genomic Map