Assay Details
Target Gene Details
Entrez Gene ID: | 207921 |
Gene Name: | family with sequence similarity 228, member B |
Gene Aliases: |
A830093I24Rik |
Location: |
Chr.12:4738397-4769267 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam228b | NM_175431.4 | 1 | 208 | NP_780640.2 |
| AK044133.1 | 1 | 209 | BAC31793.1 | |
| AK050447.1 | 1 | 174 | BAC34259.1 | |
| BC100429.1 | AAI00430.1 | |||
| BC145648.1 |
Target Gene Details
Entrez Gene ID: | 382562 |
Gene Name: | profilin family, member 4 |
Gene Aliases: |
2900024P18Rik |
Location: |
Chr.12:4768913-4778808 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pfn4 |

Genomic Map