Assay Details
Target Gene Details
Entrez Gene ID: | 23795 |
Gene Name: | anterior gradient 2 |
Gene Aliases: |
Agr2h, Gob-4, HAG-2, XAG-2, mAG-2 |
Location: |
Chr.12:35992925-36004081 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Agr2 | NM_011783.2 | 8 | 604 | NP_035913.1 |
| AB016592.1 | 8 | 603 | BAA32044.1 | |
| AF044262.1 | 8 | 604 | AAC72705.1 | |
| AK007677.1 | 8 | 623 | BAB25181.1 | |
| AK008296.1 | 7 | 672 | ||
| BC013334.1 | 8 | 622 | AAH13334.1 |
More Information
Set Membership: |
|

Genomic Map