Assay Details
Target Gene Details
Entrez Gene ID: | 72344 |
Gene Name: | ubiquitin specific peptidase 36 |
Gene Aliases: |
2700002L06Rik, mKIAA1453 |
Location: |
Chr.11:118259651-118290244 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp36 | NM_001033528.1 | NP_001028700.1 | ||
| AK028572.1 | 2 | 342 | ||
| AK049205.1 | 2 | 292 | ||
| AK079342.1 | 2 | 182 | BAC37614.1 | |
| AK129363.1 | 2 | 95 | BAC98173.1 | |
| AK165635.1 | 2 | 245 | BAE38308.1 |
More Information
Set Membership: |
|

Genomic Map