Assay Details
Target Gene Details
Entrez Gene ID: | 403205 |
Gene Name: | anterior gradient 3 |
Gene Aliases: |
BCMP11, E030025L21Rik, Gm888 |
Location: |
Chr.12:35925621-35949737 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Agr3 | NM_207531.3 | NP_997414.2 | ||
| AK053177.1 | BAC35297.1 | |||
| BC023499.1 | AAH23499.1 | |||
| BE650904.1 |
More Information
Set Membership: |
|

Genomic Map