Assay Details
Target Gene Details
Entrez Gene ID: | 70897 |
Gene Name: | family with sequence similarity 71, member D |
Gene Aliases: |
4921509E07Rik, 4930516C23Rik, GARI-L2 |
Location: |
Chr.12:78691488-78734519 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam71d | NM_027597.4 | 4 | 579 | NP_081873.2 |
| NM_029069.1 | 4 | 584 | NP_083345.1 | |
| AB473726.1 | 2 | 259 | ||
| AK014851.1 | 4 | 585 | ||
| AK015802.1 | 4 | 432 | ||
| BC051149.1 | 4 | 439 | AAH51149.1 |

Genomic Map