Assay Details
Target Gene Details
Entrez Gene ID: | 19141 |
Gene Name: | legumain |
Gene Aliases: |
AEP, AI746452, AU022324, Prsc1 |
Location: |
Chr.12:102394098-102439697 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lgmn | NM_011175.2 | NP_035305.1 | ||
| AF044266.1 | AAF21659.1 | |||
| AJ000990.1 | CAA04439.1 | |||
| AK148104.1 | ||||
| AK178161.1 | ||||
| BC132515.1 | ||||
| BC132519.1 |
More Information
Set Membership: |
|

Genomic Map